Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid, 97%

$56.96 - $56.96

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
CAS 186517-41-9
Molecular Formula C8H3ClF4O2
Formula Weight 242.56
MDL Number MFCD01631342
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2304903

  SDS
alfa aesar™
B2304903
1g Each for $56.96
Description & Specifications

Specifications

Formula Weight 242.56
Chemical Name or Material 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoic acid
Assay 97%
Molecular Formula C8H3ClF4O2
MDL Number MFCD01631342
CAS 186517-41-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 96° to 98°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.