Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
CAS 72537-17-8
Formula Weight 199.53
MDL Number MFCD00191918
UN Number UN1993
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6472303

 
alfa aesar™
H6472303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6472306

 
alfa aesar™
H6472306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6472314

 
alfa aesar™
H6472314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.53
Boiling Point 50° to 55°C (11mmHg)
Melting Point 16° to 20°C (dec.)
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
Physical Form Liquid
Assay 98%
MDL Number MFCD00191918
CAS 72537-17-8
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Refractive Index 1.4330
DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Density 1.524g/mL
Color Colorless
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.