Learn More
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6472306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine | |
16°C to 20°C (decomposition) | |
1.524 g/mL | |
52°C (126°F) | |
1.433 | |
5 g | |
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine | |
FC1=C(Cl)C=C(C=N1)C(F)(F)F | |
199.53 | |
199.53 | |
Liquid |
72537-17-8 | |
Colorless | |
50°C to 55°C (11 mmHg) | |
C6H2ClF4N | |
MFCD00191918 | |
UN1993 | |
GDSROTVTTLUHCO-UHFFFAOYSA-N | |
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine | |
1268075 | |
98% |
Chemical Identifiers
72537-17-8 | |
199.53 | |
GDSROTVTTLUHCO-UHFFFAOYSA-N | |
1268075 | |
FC1=C(Cl)C=C(C=N1)C(F)(F)F |
C6H2ClF4N | |
MFCD00191918 | |
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine | |
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures