Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)phenol, 97+%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
CAS 261763-12-6
Molecular Formula C7H3ClF4O
Formula Weight 214.55
MDL Number MFCD01631514
UN Number UN3265
Synonym 3-Chloro-4-fluoro-5-hydroxybenzotrifluoride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2336803

  SDS
alfa aesar™
B2336803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2336806

  SDS
alfa aesar™
B2336806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.55
CAS 261763-12-6
Quantity 1g
Synonym 3-Chloro-4-fluoro-5-hydroxybenzotrifluoride
Chemical Name or Material 3-Chloro-2-fluoro-5-(trifluoromethyl)phenol
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
UN Number UN3265
Health Hazard 1 Danger
Molecular Formula C7H3ClF4O
MDL Number MFCD01631514
Health Hazard 2 GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Assay ≥97%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.