Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2-cyanobenzeneboronic acid, 95%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2-cyanobenzeneboronic acid
CAS 1217500-67-8
Formula Weight 214.94
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6138903

 
alfa aesar™
H6138903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH61389MD

 
alfa aesar™
H61389MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.94
Chemical Name or Material 3-Chloro-2-cyanobenzeneboronic acid
Physical Form Solid
Assay 95%
CAS 1217500-67-8
Quantity 1g