Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-dimethylaniline hydrochloride, 99%

$118.89 - $118.89

Chemical Identifiers

Chemical Name or Material 3-Chloro-2,6-dimethylaniline hydrochloride
CAS 72725-98-5
Molecular Formula C8H10ClN·HCl
Formula Weight 192.09
MDL Number MFCD02683075
Synonym 3-Chloro-2,6-xylidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2185114

  SDS
alfa aesar™
B2185114
25g Each for $118.89
Description & Specifications

Specifications

Formula Weight 192.09
Molecular Formula C8H10ClN·HCl
MDL Number MFCD02683075
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point ≈240°C (dec.)
Health Hazard 1 Danger
Chemical Name or Material 3-Chloro-2,6-dimethylaniline hydrochloride
CAS 72725-98-5
Quantity 25g
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 3-Chloro-2,6-xylidine hydrochloride
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.