missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid, 98%

$44.73 - $142.55

Chemical Identifiers

CAS 175135-56-5
Molecular Formula C9H8ClNO6
Formula Weight 261.62
MDL Number MFCD00052955
Chemical Name or Material 3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1598803 SDS Alfa Aesar™
A1598803
1g Each for $44.73
AAA1598806 SDS Alfa Aesar™
A1598806
5g Each for $142.55
Specifications Description & Specifications

Chemical Identifiers

175135-56-5
261.62
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
C9H8ClNO6
MFCD00052955

Specifications

129° to 132°C
175135-56-5
261.62
3-Chloro-2,6-dimethoxy-5-nitrobenzoic acid
1g
C9H8ClNO6
MFCD00052955
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning