missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-difluorobenzoyl chloride, 97%

$85.14 - $293.00

Chemical Identifiers

CAS 261762-43-0
Molecular Formula C7H2Cl2F2O
Molecular Weight (g/mol) 210.989
MDL Number MFCD01631324
InChI Key LUTDLSSUGZTZBP-UHFFFAOYSA-N
Synonym 3-chloro-2,6-difluorobenzoylchloride, acmc-1cff7, 3-chloro-2,6-difluoro-benzoyl chloride, benzoyl chloride,3-chloro-2,6-difluoro, 3-chloranyl-2,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 3-chloro-2,6-difluoro-9ci
PubChem CID 2773540
IUPAC Name 3-chloro-2,6-difluorobenzoyl chloride
SMILES C1=CC(=C(C(=C1F)C(=O)Cl)F)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1824403 View Documents Alfa Aesar™
L1824403
1g Each for $85.14
 
AAL1824406 View Documents Alfa Aesar™
L1824406
5g Each for $293.00
 
Specifications

Chemical Identifiers

261762-43-0
210.989
LUTDLSSUGZTZBP-UHFFFAOYSA-N
2773540
C1=CC(=C(C(=C1F)C(=O)Cl)F)Cl
C7H2Cl2F2O
MFCD01631324
3-chloro-2,6-difluorobenzoylchloride, acmc-1cff7, 3-chloro-2,6-difluoro-benzoyl chloride, benzoyl chloride,3-chloro-2,6-difluoro, 3-chloranyl-2,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 3-chloro-2,6-difluoro-9ci
3-chloro-2,6-difluorobenzoyl chloride

Specifications

1g
C7H2Cl2F2O
UN3265
LUTDLSSUGZTZBP-UHFFFAOYSA-N
3-chloro-2,6-difluorobenzoyl chloride
2773540
97%
261762-43-0
MFCD01631324
3-chloro-2,6-difluorobenzoylchloride, acmc-1cff7, 3-chloro-2,6-difluoro-benzoyl chloride, benzoyl chloride,3-chloro-2,6-difluoro, 3-chloranyl-2,6-bis fluoranyl benzoyl chloride, benzoyl chloride, 3-chloro-2,6-difluoro-9ci
C1=CC(=C(C(=C1F)C(=O)Cl)F)Cl
210.989
211
3-Chloro-2,6-difluorobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.