Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-difluorobenzonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3-Chloro-2,6-difluorobenzonitrile
CAS 86225-73-2
Molecular Formula C7H2ClF2N
Formula Weight 173.55
MDL Number MFCD06660172
UN Number UN3276
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2633503

  SDS
alfa aesar™
H2633503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2633506

  SDS
alfa aesar™
H2633506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2633514

  SDS
alfa aesar™
H2633514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 173.55
CAS 86225-73-2
MDL Number MFCD06660172
Molecular Formula C7H2ClF2N
Health Hazard 1 Danger
Chemical Name or Material 3-Chloro-2,6-difluorobenzonitrile
UN Number UN3276
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
Quantity 1g
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

Applications
3-Chloro-2,6-difluorobenzonitrile is used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.