Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-difluorobenzoic acid, 97%

$59.38 - $387.24

Chemical Identifiers

Chemical Name or Material 3-Chloro-2,6-difluorobenzoic acid
CAS 225104-76-7
Molecular Formula C7H3ClF2O2
Formula Weight 192.55
MDL Number MFCD01631322
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2389303

  SDS
alfa aesar™
B2389303
1g Each for $59.38

AAB2389306

  SDS
alfa aesar™
B2389306
5g Each for $218.49

AAB2389309

  SDS
alfa aesar™
B2389309
10g Each for $387.24
Description & Specifications

Specifications

Formula Weight 192.55
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C7H3ClF2O2
Melting Point 126° to 128°C
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 225104-76-7
MDL Number MFCD01631322
Chemical Name or Material 3-Chloro-2,6-difluorobenzoic acid
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.