Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,6-difluorobenzamide, 97+%

$174.78 - $174.78

Chemical Identifiers

Chemical Name or Material 3-Chloro-2,6-difluorobenzamide
CAS 261762-41-8
Molecular Formula C7H4ClF2NO
Formula Weight 191.56
MDL Number MFCD01631386
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2406506

  SDS
alfa aesar™
B2406506
5g Each for $174.78
Description & Specifications

Specifications

Formula Weight 191.56
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥97%
Health Hazard 1 Warning
CAS 261762-41-8
Melting Point 133° to 136°C
MDL Number MFCD01631386
Quantity 5g
Chemical Name or Material 3-Chloro-2,6-difluorobenzamide
Molecular Formula C7H4ClF2NO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.