Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Chloro-2',6'-difluoroacetophenone, 97+%

$92.97 - $326.97

Chemical Identifiers

Chemical Name or Material 3'-Chloro-2',6'-difluoroacetophenone
CAS 177942-50-6
Molecular Formula C8H5ClF2O
Formula Weight 190.58
MDL Number MFCD01631329
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2444503

  SDS
alfa aesar™
B2444503
1g Each for $92.97

AAB2444506

  SDS
alfa aesar™
B2444506
5g Each for $326.97
Description & Specifications

Specifications

Formula Weight 190.58
MDL Number MFCD01631329
Quantity 1g
Health Hazard 1 Warning
CAS 177942-50-6
Assay ≥97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 3'-Chloro-2',6'-difluoroacetophenone
Molecular Formula C8H5ClF2O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
It is an intermediate in organic syntheses.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.