missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-2,4-pentanedione, 98%, ACROS Organics™

$97.61 - $97.61

Chemical Identifiers

CAS 1694-29-7
Molecular Formula C5H7ClO2
Molecular Weight (g/mol) 134.559
MDL Number MFCD00009651
InChI Key VLRGXXKFHVJQOL-UHFFFAOYSA-N
Synonym 3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
PubChem CID 74328
IUPAC Name 3-chloropentane-2,4-dione
SMILES CC(=O)C(C(=O)C)Cl
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC210670025 SDS View Documents Acros Organics
AC210670025
2.5L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210675000 SDS View Documents Acros Organics
AC210675000
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210670050 SDS View Documents Acros Organics
210670050
5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC210671000 SDS View Documents Acros Organics
210671000
100mL Each for $97.61
Specifications

Chemical Identifiers

1694-29-7
134.559
VLRGXXKFHVJQOL-UHFFFAOYSA-N
74328
CC(=O)C(C(=O)C)Cl
C5H7ClO2
MFCD00009651
3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
3-chloropentane-2,4-dione

Specifications

1.179g/mL
Glass bottle
Brown to Yellow
1694-29-7
100.0
C5H7ClO2
MFCD00009651
3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc
CC(=O)C(C(=O)C)Cl
134.559
134.56
≥97.5%
Authentic
1.4825 to 1.4845 (20°C, 589nm)
2.5L
97.5
98%
CH3COCH(Cl)COCH3
01, 785
VLRGXXKFHVJQOL-UHFFFAOYSA-N
3-chloropentane-2,4-dione
74328
Liquid
3-Chloro-2, 4-pentanedione, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 216-902-4

TSCA : TSCA