Learn More
3-Chloro-2,4-pentanedione, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 210671000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Chloro-2, 4-pentanedione | |
1694-29-7 | |
100.0 | |
Brown to Yellow | |
49°C to 52°C (18.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4825 to 1.4845 | |
MFCD00009651 | |
01, 785 | |
3-chloro-2,4-pentanedione, 3-chloroacetylacetone, 2,4-pentanedione, 3-chloro, 1-acetyl-1-chloroacetone, 3-chloro 2,4-pentanedione, 3-chloro.2,4-pentanedione, 3-chloro-pentane-2,4-dione, 2,4-pentanedione,3-chloro, 3-chloro-2,4-pentanedione, produced by wacker chemie ag, burghausen, germany gc | |
VLRGXXKFHVJQOL-UHFFFAOYSA-N | |
3-chloropentane-2,4-dione | |
74328 | |
98% |
98% | |
97.5 | |
-15°C | |
1.1790g/mL | |
56°C | |
97.5% min. (GC) | |
C5H7ClO2 | |
CH3COCH(Cl)COCH3 | |
100 mL | |
1.179 | |
Solubility in water: 7.65g/L (20°C). Other solubilities: soluble in organic solvents | |
CC(=O)C(C(=O)C)Cl | |
134.56 | |
134.56 | |
Liquid |
Chemical Identifiers
1694-29-7 | |
134.56 | |
VLRGXXKFHVJQOL-UHFFFAOYSA-N | |
3-chloropentane-2,4-dione |
C5H7ClO2 | |
MFCD00009651 | |
74328 | |
CC(=O)C(C(=O)C)Cl |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
GHS Signal Word: Warning
EINECSNumber : 216-902-4
TSCA : TSCA
RUO – Research Use Only