Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,4-difluorobenzaldehyde, 97%

$90.05 - $797.35

Chemical Identifiers

Chemical Name or Material 3-Chloro-2,4-difluorobenzaldehyde
CAS 127675-46-1
Molecular Formula C7H3ClF2O
Formula Weight 176.55
MDL Number MFCD04116001
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2638303

 
alfa aesar™
H2638303
1g Each for $90.05

AAH2638306

 
alfa aesar™
H2638306
5g Each for $267.84

AAH2638314

 
alfa aesar™
H2638314
25g Each for $797.35
Description & Specifications

Specifications

Formula Weight 176.55
Chemical Name or Material 3-Chloro-2,4-difluorobenzaldehyde
Assay 97%
Molecular Formula C7H3ClF2O
MDL Number MFCD04116001
CAS 127675-46-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g

Applications
3-Chloro-2,4-difluorobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.