missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-2,4,5-trifluorobenzoic acid, 97%

$26.22 - $104.41

Chemical Identifiers

CAS 101513-77-3
Molecular Formula C7H2ClF3O2
Molecular Weight (g/mol) 210.536
MDL Number MFCD00153101
InChI Key KBESHLYCSZINAJ-UHFFFAOYSA-N
Synonym 2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
PubChem CID 2734269
IUPAC Name 3-chloro-2,4,5-trifluorobenzoic acid
SMILES C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6011303 View Documents Alfa Aesar™
H6011303
1g Each for $26.22
 
AAH6011306 View Documents Alfa Aesar™
H6011306
5g Each for $104.41
 
Specifications

Chemical Identifiers

101513-77-3
210.536
KBESHLYCSZINAJ-UHFFFAOYSA-N
2734269
C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O
C7H2ClF3O2
MFCD00153101
2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
3-chloro-2,4,5-trifluorobenzoic acid

Specifications

118°C (244°F)
112°C to 116°C
101513-77-3
MFCD00153101
KBESHLYCSZINAJ-UHFFFAOYSA-N
3-chloro-2,4,5-trifluorobenzoic acid
2734269
97%
272°C
1g
C7H2ClF3O2
2,4,5-trifluoro-3-chlorobenzoic acid, 3-chloro-2,4,5-trifluoro-benzoic acid, 3-chloro-2,4,5-trifluorobenzoicacid, benzoic acid, 3-chloro-2,4,5-trifluoro, pubchem1376, acmc-1bvtw, ksc502q6n, 3-chloro-2,4,5trifluorobenzoic acid, 3-chloro-2,4,5-trilfuorobenzoic acid, 3-carboxy-2,5,6-trifluorochlorobenzene
C1=C(C(=C(C(=C1F)F)Cl)F)C(=O)O
210.536
210.54
3-Chloro-2,4,5-trifluorobenzoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning