missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Chloro-1H-indazole, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 263740050

 View more versions of this product

Catalog No. AC263740050



Specifications

Specifications

3-Chloro-1H-indazole
Glass bottle
148°C to 150°C
99%
98.5
99%
MFCD00067527
QPHAGNNWDZSKJH-UHFFFAOYSA-N
3-chloro-2H-indazole
95894
Fluffy Powder
Authentic
Beige to White
5g
29110-74-5
100.0
C7H5ClN2
1h-indazole, 3-chloro, 1h-indazole,3-chloro, 2h-indazole, 3-chloro, 3-chloro-1h-indazole, 3-chloroindazole, acmc-1cio6, ksc205k5h
C1=CC2=C(NN=C2C=C1)Cl
152.58
152.58
99%

Chemical Identifiers

29110-74-5
152.58
QPHAGNNWDZSKJH-UHFFFAOYSA-N
95894
C1=CC2=C(NN=C2C=C1)Cl
C7H5ClN2
MFCD00067527
1h-indazole, 3-chloro, 1h-indazole,3-chloro, 2h-indazole, 3-chloro, 3-chloro-1h-indazole, 3-chloroindazole, acmc-1cio6, ksc205k5h
3-chloro-2H-indazole
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 249-444-9

TSCA : TSCA

SDS Documents