Please login to your online account to display your discounted pricing

3-Chloro-1H-indazole, 99%, ACROS Organics™

$109.63 - $414.20

Chemical Identifiers

Chemical Name or Material 3-Chloro-1H-indazole
Name Note 99%
CAS 29110-74-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H5ClN2
Formula Weight 152.58
MDL Number MFCD00067527
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC263740250

  SDS
thermo fisher scientific
263740250
Glass bottle 25g Each for $414.20

AC263740050

  SDS
thermo fisher scientific
263740050
Glass bottle 5g Each for $109.63
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Fluffy Powder
Formula Weight 152.58
Quantity 25g
Color Almost white to light beige
EINECS Number 249-444-9
TSCA TSCA
CAS Max % 100.0
Packaging Glass bottle
Chemical Name or Material 3-Chloro-1H-indazole
Name Note 99%
Assay Percent Range 99%
Molecular Formula C7H5ClN2
CAS 29110-74-5
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
MDL Number MFCD00067527
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.