missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-1-phenyl-1-propyne, 97%

$41.35 - $540.32

Chemical Identifiers

CAS 3355-31-5
Molecular Formula C9H7Cl
Molecular Weight (g/mol) 150.605
MDL Number MFCD06411085
InChI Key RMPSZEZJKSUNKR-UHFFFAOYSA-N
Synonym 1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578
PubChem CID 287343
IUPAC Name 3-chloroprop-1-ynylbenzene
SMILES C1=CC=C(C=C1)C#CCCl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5351003
View Documents
Alfa Aesar™
H5351003
1g
Each for $41.35
 
AAH5351006
View Documents
Alfa Aesar™
H5351006
5g
Each for $140.51
 
AAH5351014
View Documents
Alfa Aesar™
H5351014
25g
Each for $540.32
 
Description

Description

Applications
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

3355-31-5
150.605
RMPSZEZJKSUNKR-UHFFFAOYSA-N
287343
C1=CC=C(C=C1)C#CCCl
C9H7Cl
MFCD06411085
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578
3-chloroprop-1-ynylbenzene

Specifications

1.095
1.585
3355-31-5
C6H4C≡CCH2Cl
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578
C1=CC=C(C=C1)C#CCCl
150.605
150.61
3-Chloro-1-phenyl-1-propyne
104°C (219°F)
1g
C9H7Cl
MFCD06411085
RMPSZEZJKSUNKR-UHFFFAOYSA-N
3-chloroprop-1-ynylbenzene
287343
97%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS