missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ 3-Chloro-1-phenyl-1-propyne, 97%
Manufacturer: Alfa Aesar™ H5351006
Description
Applications3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
3-Chloro-1-phenyl-1-propyne | |
104°C (219°F) | |
5g | |
C9H7Cl | |
MFCD06411085 | |
RMPSZEZJKSUNKR-UHFFFAOYSA-N | |
3-chloroprop-1-ynylbenzene | |
287343 | |
97% |
1.095 | |
1.585 | |
3355-31-5 | |
C6H4C≡CCH2Cl | |
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578 | |
C1=CC=C(C=C1)C#CCCl | |
150.605 | |
150.61 |
Chemical Identifiers
3355-31-5 | |
150.605 | |
RMPSZEZJKSUNKR-UHFFFAOYSA-N | |
287343 | |
C1=CC=C(C=C1)C#CCCl |
C9H7Cl | |
MFCD06411085 | |
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578 | |
3-chloroprop-1-ynylbenzene |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures