missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Chloro-1-phenyl-1-propyne, 97%

Manufacturer:  Alfa Aesar™ H5351003

 View more versions of this product

Catalog No. AAH5351003



Description

Description

Applications
3-Chloro-1-phenyl-1-propyne is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Chloro-1-phenyl-1-propyne
104°C (219°F)
1g
C9H7Cl
MFCD06411085
RMPSZEZJKSUNKR-UHFFFAOYSA-N
3-chloroprop-1-ynylbenzene
287343
97%
1.095
1.585
3355-31-5
C6H4C≡CCH2Cl
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578
C1=CC=C(C=C1)C#CCCl
150.605
150.61

Chemical Identifiers

3355-31-5
150.605
RMPSZEZJKSUNKR-UHFFFAOYSA-N
287343
C1=CC=C(C=C1)C#CCCl
C9H7Cl
MFCD06411085
1-chloro-3-phenyl-2-propyne, 1-phenyl-3-chloro-1-propyn, 1-phenyl-3-chloro-1-propyne, 3-chloro-1-phenyl-1-propyne, 3-chloro-l-propynyl benzene, 3-chloroprop-1-yn-1-yl benzene, 3-phenyl-2-propynyl chloride, 3-phenylpropargyl chloride, benzene, 3-chloro-1-propynyl, pubchem13578
3-chloroprop-1-ynylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents