Please login to your online account to display your discounted pricing

3-Chloro-1-methoxy-2-propanol +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Chloro-1-methoxy-2-propanol
CAS 4151-97-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H9ClO2
Linear Formula ClCH2CH(OH)CH2OCH3
Formula Weight 124.57
MDL Number MFCD00018935
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC392315000

  SDS
thermo fisher scientific
AC392315000
Glass bottle 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Formula Weight 124.57
Physical Form Oily Liquid
Quantity 500mL
Color Clear faintly yellow
Density 1.16g/mL
CAS Max % 100.0
CAS Min % 98.0
Linear Formula ClCH2CH(OH)CH2OCH3
Refractive Index 1.4460 to 1.4480 (20°C, 589nm)
Packaging Glass bottle
Molecular Formula C4H9ClO2
Assay Percent Range 98+%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00018935
Infrared Spectrum Authentic
CAS 4151-97-7
Chemical Name or Material 3-Chloro-1-methoxy-2-propanol