Learn More
3-Chloro-1,2-propanediol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 148120250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Chloro-1, 2-propanediol | |
98.5 | |
-40.0°C | |
1.3220g/mL | |
135°C | |
98.5% min. (GC) | |
C3H7ClO2 | |
ClCH2CH(OH)CH2OH | |
25 mL | |
15, 2146 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,methanol and ether,slightly soluble in toluene | |
OCC(O)CCl | |
110.54 | |
215 mPa.s (20°C) | |
110.54 | |
Liquid |
96-24-2 | |
100.0 | |
Yellow | |
213.0°C | |
Authentic | |
Glass bottle | |
1.4790 to 1.4810 | |
MFCD00004712 | |
1.322 | |
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
3-chloropropane-1,2-diol | |
7290 | |
CHEBI:18721 | |
99% |
Chemical Identifiers
96-24-2 | |
110.54 | |
SSZWWUDQMAHNAQ-UHFFFAOYNA-N | |
7290 | |
3-chloropropane-1,2-diol |
C3H7ClO2 | |
MFCD00004712 | |
3-chloro-1,2-propanediol, alpha-chlorohydrin, chlorodeoxyglycerol, glycerol alpha-monochlorohydrin, epibloc, 1,2-propanediol, 3-chloro, a-chlorohydrin, 3-chloropropanediol, 3-chloropropylene glycol, glycerol chlorohydrin | |
CHEBI:18721 | |
OCC(O)CCl |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Fatal if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Fatal if inhaled.
May cause respiratory irritation.
Causes skin irritation.<br
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
GHS Signal Word: Danger
EINECSNumber : 202-492-4
RUO – Research Use Only