missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Carboxybenzeneboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6419506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Carboxybenzeneboronic acid pinacol ester | |
206°C to 210°C (decomposition) | |
C13H17BO4 | |
5 g | |
OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid | |
2734653 | |
97% |
269409-73-6 | |
White | |
MFCD03411930 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044 | |
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O | |
248.09 | |
248.09 | |
Powder |
Chemical Identifiers
269409-73-6 | |
248.09 | |
OPWAPCOSDAFWFB-UHFFFAOYSA-N | |
2734653 | |
CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C(O)=O |
C13H17BO4 | |
MFCD03411930 | |
3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronic acid pinacol ester, 3-carboxyphenylboronic acid, pinacol ester, 3-carboxyphenyl boronic acid, pinacol ester, benzoic acid, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxyphenylboronicacidpinacolester, pubchem17048, 3-cpbe, amtb044 | |
3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only