Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carboxybenzeneboronic acid

$186.84 - $186.84

Chemical Identifiers

Chemical Name or Material 3-Carboxybenzeneboronic acid
CAS 25487-66-5
Molecular Formula C7H7BO4
Formula Weight 165.94
MDL Number MFCD00036833
Beilstein 2967400
Synonym 3-Boronobenzoic acid; 3-Carboxyphenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2531506

  SDS
alfa aesar™
B2531506
5g Each for $186.84
Description & Specifications

Specifications

Formula Weight 165.94
Chemical Name or Material 3-Carboxybenzeneboronic acid
Molecular Formula C7H7BO4
MDL Number MFCD00036833
CAS 25487-66-5
Beilstein 2967400
Synonym 3-Boronobenzoic acid; 3-Carboxyphenylboronic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CY8750000
Melting Point ≈240°C (dec.)
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.