Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carboxybenzaldehyde, 97%

$89.80 - $293.82

Chemical Identifiers

Chemical Name or Material 3-Carboxybenzaldehyde
CAS 619-21-6
Molecular Formula C8H6O3
Formula Weight 150.13
MDL Number MFCD00039575
Beilstein 2206413
Synonym 3-Formylbenzoic acid; Isophthalaldehydic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2127703

  SDS
alfa aesar™
B2127703
1g Each for $89.80

AAB2127706

  SDS
alfa aesar™
B2127706
5g Each for $293.82
Description & Specifications

Specifications

Formula Weight 150.13
Chemical Name or Material 3-Carboxybenzaldehyde
TSCA TSCA
Assay 97%
Molecular Formula C8H6O3
MDL Number MFCD00039575
CAS 619-21-6
Beilstein 2206413
Synonym 3-Formylbenzoic acid; Isophthalaldehydic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 173° to 175°C
Quantity 1g

Applications
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

Solubility
Soluble in methanol (100 mg/mL) gives a clear to slightly hazy, colorless to very faintly brown.

Notes
Air sensitive. Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.