Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester
CAS 882679-10-9
Formula Weight 266.07
MDL Number MFCD12546519
Synonym 3-Carboxy-4-fluorophenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH62683MD

 
alfa aesar™
H62683MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6268303

 
alfa aesar™
H6268303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.07
Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
MDL Number MFCD12546519
CAS 882679-10-9
Synonym 3-Carboxy-4-fluorophenylboronic acid pinacol ester
Quantity 250mg