Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carboxy-4-fluorobenzeneboronic acid, 97%

$33.30 - $233.55

Chemical Identifiers

Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid
CAS 872460-12-3
Molecular Formula C7H6BFO4
Formula Weight 183.93
MDL Number MFCD03095129
Synonym 5-Borono-2-fluorobenzoic acid; 3-Carboxy-4-fluorophenylboronic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2831303

  SDS
alfa aesar™
H2831303
1g Each for $33.30

AAH2831309

  SDS
alfa aesar™
H2831309
10g Each for $233.55
Description & Specifications

Specifications

Formula Weight 183.93
CAS 872460-12-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 217° to 219°C
Quantity 1g
Synonym 5-Borono-2-fluorobenzoic acid; 3-Carboxy-4-fluorophenylboronic acid
MDL Number MFCD03095129
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid
Molecular Formula C7H6BFO4
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.