missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Carbamyl-1-methylpyridinium Chloride, TCI America™

Manufacturer:  TCI America M03755G

 View more versions of this product

Catalog No. M03755G



Specifications

Specifications

3-Carbamyl-1-methylpyridinium Chloride
5g
C7H9ClN2O
3-Carbamoyl-1-methylpyridinium Chloride
C[N+]1=CC=CC(=C1)C(=O)N.[Cl-]
172.612
172.61
≥98.0% (HPLC,T)
White-Yellow
1005-24-9
MFCD00060042
BWVDQVQUNNBTLK-UHFFFAOYSA-N
1-methylpyridin-1-ium-3-carboxamide;chloride
70495
Crystal-Powder at 20°C

Chemical Identifiers

1005-24-9
172.612
BWVDQVQUNNBTLK-UHFFFAOYSA-N
70495
C[N+]1=CC=CC(=C1)C(=O)N.[Cl-]
C7H9ClN2O
MFCD00060042
3-Carbamoyl-1-methylpyridinium Chloride
1-methylpyridin-1-ium-3-carboxamide;chloride
Safety and Handling

Safety and Handling

RTECSNumber : UU1925000

TSCA : No

Documents