missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6213003

 View more versions of this product

Catalog No. AAH6213003

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Carbamoyl-4-fluorobenzeneboronic acid pinacol ester
1374508-59-4
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)N
265.091
265.09
96%
1g
C13H17BFNO3
ZETAUVYXFMYTIN-UHFFFAOYSA-N
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
90376049
Solid

Chemical Identifiers

1374508-59-4
265.091
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzamide
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
C13H17BFNO3
ZETAUVYXFMYTIN-UHFFFAOYSA-N
90376049
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)N
Documents