missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Carbamoyl-4-fluorobenzeneboronic acid, 97%

Manufacturer:  Alfa Aesar™ H5310203

 View more versions of this product

Catalog No. AAH5310203



Specifications

Specifications

3-Carbamoyl-4-fluorobenzeneboronic acid
1g
C7H7BFNO3
3-carbamoyl-4-fluorophenyl boronic acid, 3-carbamoyl-4-fluorobenzeneboronic acid, 3-aminocarbonyl-4-fluorophenylboronic acid, 3-aminocarbonyl-4-fluorobenzeneboronic acid, acmc-209qky, 3-carbamoyl-4-fluorophenyl boronicacid, 4-fluoro-3-carbamoyl phenylboronic acid, 3-aminocarbonyl-4-fluoranyl-phenyl boronic acid, boronic acid,b-3-aminocarbonyl-4-fluorophenyl
B(C1=CC(=C(C=C1)F)C(=O)N)(O)O
182.945
182.95
257°C to 259°C
874219-34-8
MFCD08235053
RNNYXSWJFLHIRC-UHFFFAOYSA-N
(3-carbamoyl-4-fluorophenyl)boronic acid
44717530
97%

Chemical Identifiers

874219-34-8
182.945
RNNYXSWJFLHIRC-UHFFFAOYSA-N
44717530
B(C1=CC(=C(C=C1)F)C(=O)N)(O)O
C7H7BFNO3
MFCD08235053
3-carbamoyl-4-fluorophenyl boronic acid, 3-carbamoyl-4-fluorobenzeneboronic acid, 3-aminocarbonyl-4-fluorophenylboronic acid, 3-aminocarbonyl-4-fluorobenzeneboronic acid, acmc-209qky, 3-carbamoyl-4-fluorophenyl boronicacid, 4-fluoro-3-carbamoyl phenylboronic acid, 3-aminocarbonyl-4-fluoranyl-phenyl boronic acid, boronic acid,b-3-aminocarbonyl-4-fluorophenyl
(3-carbamoyl-4-fluorophenyl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents