Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Butyrylcoumarin, 98+%

$47.46 - $661.47

Chemical Identifiers

Chemical Name or Material 3-Butyrylcoumarin
CAS 1846-73-7
Molecular Formula C13H12O3
Formula Weight 216.24
MDL Number MFCD00052893
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1526203

  SDS
alfa aesar™
A1526203
1g Each for $47.46

AAA1526206

  SDS
alfa aesar™
A1526206
5g Each for $159.72

AAA1526214

  SDS
alfa aesar™
A1526214
25g Each for $661.47
Description & Specifications

Specifications

Formula Weight 216.24
Chemical Name or Material 3-Butyrylcoumarin
Assay ≥98%
Molecular Formula C13H12O3
MDL Number MFCD00052893
CAS 1846-73-7
Melting Point 92° to 96°C
Quantity 1g