Please login to your online account to display your discounted pricing

(3-Butynyloxy)trimethylsilane 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (3-Butynyloxy)trimethylsilane
CAS 17869-75-9
CAS Min % 97.5
CAS Max % 100.0
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H14OSi
Linear Formula (CH3)3SiOCH2CH2CCH
Formula Weight 142.27g/mol
MDL Number MFCD00190214
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC390560010

  SDS
thermo fisher scientific
390560010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4110 to 1.4130 (20°C, 589nm)
Density 0.8330g/mL
Color Clear slightly yellow
Quantity 1g
Chemical Name or Material (3-Butynyloxy)trimethylsilane
CAS 17869-75-9
CAS Min % 97.5
CAS Max % 100.0
Physical Form Clear slightly yellow liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C7H14OSi
Linear Formula (CH3)3SiOCH2CH2CCH
Formula Weight 142.27g/mol
MDL Number MFCD00190214
Synonym 1-Trimethylsilyloxy-3-butyne
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.