missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Butyn-2-one, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 330440025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Butyn-2-one | |
0.8700g/mL | |
−1°C | |
95% min. (GC) | |
C4H4O | |
HC≡CCOCH3 | |
2.5g | |
0.87 | |
XRGPFNGLRSIPSA-UHFFFAOYSA-N | |
but-3-yn-2-one | |
15018 | |
68.06 |
1423-60-5 | |
85.0°C | |
Authentic | |
Glass bottle | |
1.4045 to 1.4095 | |
MFCD00008775 | |
03,38 | |
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch | |
CC(=O)C#C | |
68.08 | |
CHEBI:48060 | |
96% |
Chemical Identifiers
1423-60-5 | |
68.08 | |
XRGPFNGLRSIPSA-UHFFFAOYSA-N | |
15018 | |
but-3-yn-2-one |
C4H4O | |
MFCD00008775 | |
3-butyn-2-one, 1-butyn-3-one, methyl ethynyl ketone, acetylacetylene, butyn-2-one, acetylethyne, 1-butyne-3-one, 3-butyne-2-one, ethynyl methyl ketone, ch3coc#ch | |
CHEBI:48060 | |
CC(=O)C#C |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 215-834-2
RUO â Research Use Only