Please login to your online account to display your discounted pricing

3-Butyn-2-one, 96%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  330440100

 View more versions of this product

Catalog No. AC330440100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95% (GC)
Formula Weight 68.06
Physical Form Liquid
Quantity 10g
Color Clear yellow to orange-brown
Density 0.87g/mL
Linear Formula HC~2CCOCH3
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 3-Butyn-2-one
Refractive Index 1.4045 to 1.4095 (20°C, 589nm)
Assay Percent Range 96%
Molecular Formula C4H4O
CAS 1423-60-5

Specifications

Chemical Name or Material 3-Butyn-2-one
Assay ≥95% (GC)
Assay Percent Range 96%
Molecular Formula C4H4O
Linear Formula HC~2CCOCH3
Formula Weight 68.06