missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Butyn-2-ol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC245310100

 View more versions of this product

Catalog No. AC245310100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Butyn-2-ol
26°C
Glass bottle
0.89
Colorless to Yellow
2028-63-9
100.0
C4H6O
1-butyn-3-ol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, 1-methylpropargyl alcohol, 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol, methylethynylcarbinol
GKPOMITUDGXOSB-UHFFFAOYSA-N
but-3-yn-2-ol
16239
Liquid
0.8900g/mL
Authentic
1.4250 to 1.427
108°C to 111°C
10g
96.0
96% min. (GC)
MFCD00004541
Solubility in water: miscible. Other solubilities: miscible with methanol
CC(C#C)O
70.09
70.09
97%

Chemical Identifiers

2028-63-9
70.09
GKPOMITUDGXOSB-UHFFFAOYSA-N
16239
CC(C#C)O
C4H6O
MFCD00004541
1-butyn-3-ol, 1-ethynylethanol, 1-methyl-2-propynyl alcohol, 1-methylpropargyl alcohol, 3-butyn-2-ol, 3-butyne-2-ol, 3-hydroxy-1-butyne, alpha-methylpropargyl alcohol, dl-3-butyn-2-ol, methylethynylcarbinol
but-3-yn-2-ol
Safety and Handling

Safety and Handling

GHS H Statement
Fatal in contact with skin.
May cause respiratory irritation.
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Fatal if inhaled.

GHS P Statement
If exposed or you feel unwell: Call a POISON CENTRE or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash wi

GHS Signal Word: Danger

EINECSNumber : 217-978-1

SDS Documents