missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Butyn-1-ol 97%, ACROS Organics™

Manufacturer:  Acros Organics 157330050

 View more versions of this product

Catalog No. AC157330050



Specifications

Specifications

3-Butyn-1-ol
36°C
Glass bottle
128.8°C
-63.6°C
97%
96.0
97%
HC≡CCH2CH2OH
01, IV, 2219
OTJZCIYGRUNXTP-UHFFFAOYSA-N
but-3-yn-1-ol
13566
70.09
97%
0.9260g/mL
Authentic
1.4397 to 1.4417
Colorless to Yellow
5mL
927-74-2
100.0
C4H6O
MFCD00002955
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
C#CCCO
70.091
CHEBI:27444
Liquid

Chemical Identifiers

927-74-2
70.091
OTJZCIYGRUNXTP-UHFFFAOYSA-N
13566
but-3-yn-1-ol
C4H6O
MFCD00002955
3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
CHEBI:27444
C#CCCO
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 213-161-9

RTECSNumber : ES0710000

TSCA : TSCA

SDS Documents