missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Butylthiophene 98.0+%, TCI America™

$47.00 - $160.69

Chemical Identifiers

CAS 34722-01-5
Molecular Formula C8H12S
Molecular Weight (g/mol) 140.244
MDL Number MFCD00143179
InChI Key KPOCSQCZXMATFR-UHFFFAOYSA-N
Synonym 3-butyl thiophene
PubChem CID 566339
IUPAC Name 3-butylthiophene
SMILES CCCCC1=CSC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B14581G View Documents Tci America
B14581G
1g Each for $47.00
 
B14585G View Documents Tci America
B14585G
5g Each for $160.69
 
Specifications

Chemical Identifiers

34722-01-5
140.244
KPOCSQCZXMATFR-UHFFFAOYSA-N
566339
CCCCC1=CSC=C1
C8H12S
MFCD00143179
3-butyl thiophene
3-butylthiophene

Specifications

183°C
1g
C8H12S
1993
KPOCSQCZXMATFR-UHFFFAOYSA-N
3-butylthiophene
566339
Clear Liquid at 20°C
3-Butylthiophene
Yellow
34722-01-5
MFCD00143179
3-butyl thiophene
CCCCC1=CSC=C1
140.244
140.24
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No