Please login to your online account to display your discounted pricing

3-Butenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, ACROS Organics™


Quantity 100mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC427461000

 View more versions of this product

Catalog No. AC427461000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 159.31
Physical Form Solution
Quantity 100mL
Color Gray to brown
Packaging AcroSeal™ Glass Bottle
Density 0.9370g/mL
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Health Hazard 3 GHS P Statement Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Chemical Name or Material 3-Butenylmagnesium bromide
Name Note 0.5M solution in THF
Molecular Formula C4H7BrMg
MDL Number MFCD00209554
CAS 7103-09-5
Health Hazard 1 Danger
CAS Max % 10.2
CAS Min % 89.8
Linear Formula H2C=CHCH2CH2MgBr
CAS Min % 6.8
CAS 109-99-9
CAS Max % 93.2

Specifications

Chemical Name or Material 3-Butenylmagnesium bromide
Name Note 0.5M solution in THF
CAS 7103-09-5
CAS Min % 6.8
CAS Max % 10.2
CAS 109-99-9
CAS Min % 89.8
CAS Max % 93.2
Molecular Formula C4H7BrMg
Linear Formula H2C=CHCH2CH2MgBr
Formula Weight 159.31
MDL Number MFCD00209554
Accessories

Danger

GHS P Statement Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.

SDS & Safety Documents