missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Butenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC427468000

 View more versions of this product

Catalog No. AC427468000



Description

Description

Specifications

Specifications

3-Butenylmagnesium bromide
-30°C
0.937
Brown to Gray
0.5M solution in THF
6.8,89.8
C4H7BrMg
MFCD00209554
ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
131675247
Solution
0.9370g/mL
AcroSeal™ Glass Bottle
65°C
800mL
7103-09-5,109-99-9
10.2,93.2
H2C=CHCH2CH2MgBr
grignard reagent
CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
159.31
159.31

Chemical Identifiers

7103-09-5
159.31
ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
131675247
CC[N+](C)(C)C1=CC=C(C=C1)C(=C2C=CC(=[N+](C)C)C=C2)C3=CC=C(C=C3)N(C)C.[Cl-].[Cl-].[Cl-].[Zn+2].[Br-]
C4H7BrMg
MFCD00209554
grignard reagent
zinc;[4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]phenyl]-ethyl-dimethylazanium;bromide;trichloride
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
Highly flammable liquid and vapor.
Suspected of causing cancer.
Reacts violently

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS Documents