Learn More
3-Buten-1-amine, tech. 90%, Thermo Scientific Chemicals
$247.14 - $995.48
Chemical Identifiers
CAS | 2524-49-4 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.12 |
MDL Number | MFCD03425859 |
InChI Key | ASVKKRLMJCWVQF-UHFFFAOYSA-N |
Synonym | 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene |
PubChem CID | 443732 |
ChEBI | CHEBI:31108 |
IUPAC Name | but-3-en-1-amine |
SMILES | NCCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2015703
|
Thermo Scientific Chemicals
L2015703 |
1 g |
Each for $247.14
|
|
|||||
AAL2015706
|
Thermo Scientific Chemicals
L2015706 |
5 g |
Each for $995.48
|
|
|||||
Description
3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimerâs disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimerâs disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong oxidizing agents and strong bases.
Chemical Identifiers
2524-49-4 | |
71.12 | |
ASVKKRLMJCWVQF-UHFFFAOYSA-N | |
443732 | |
but-3-en-1-amine |
C4H9N | |
MFCD03425859 | |
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene | |
CHEBI:31108 | |
NCCC=C |
Specifications
2524-49-4 | |
74°C to 77°C | |
Amine-like | |
1.424 | |
1 g | |
1735803 | |
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene | |
ASVKKRLMJCWVQF-UHFFFAOYSA-N | |
but-3-en-1-amine | |
443732 | |
71.12 | |
3-Buten-1-amine |
0.777 | |
−5°C (23°F) | |
C4H9N | |
MFCD03425859 | |
UN2733 | |
Air Sensitive | |
Miscible with water. | |
NCCC=C | |
71.12 | |
CHEBI:31108 | |
97% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only