Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromoveratrole, 95%

$266.70 - $604.22

Chemical Identifiers

Chemical Name or Material 3-Bromoveratrole
CAS 5424-43-1
Molecular Formula C8H9BrO2
Formula Weight 217.06
MDL Number MFCD00185638
Synonym 1-Bromo-2,3-dimethoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2783506

  SDS
alfa aesar™
H2783506
5g Each for $604.22

AAH2783503

  SDS
alfa aesar™
H2783503
1g Each for $266.70
Description & Specifications

Specifications

Formula Weight 217.06
Molecular Formula C8H9BrO2
Quantity 5g
CAS 5424-43-1
MDL Number MFCD00185638
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Chemical Name or Material 3-Bromoveratrole
Synonym 1-Bromo-2,3-dimethoxybenzene
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.