Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromothiophenol, 97%

$44.32 - $526.43

Chemical Identifiers

Chemical Name or Material 3-Bromothiophenol
CAS 6320-01-0
Molecular Formula C6H5BrS
Formula Weight 189.08
MDL Number MFCD00009603
Beilstein 1926242
Synonym 3-Bromobenzenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2509203

 
alfa aesar™
B2509203
1g Each for $44.32

AAB2509206

 
alfa aesar™
B2509206
5g Each for $134.54

AAB2509214

 
alfa aesar™
B2509214
25g Each for $526.43
Description & Specifications

Specifications

Formula Weight 189.08
Chemical Name or Material 3-Bromothiophenol
Assay 97%
Molecular Formula C6H5BrS
MDL Number MFCD00009603
CAS 6320-01-0
Beilstein 1926242
Synonym 3-Bromobenzenethiol
Boiling Point 119° to 121°C (21mmHg)
Quantity 1g
Flash Point 105°C (221°F)
Odor Unpleasant
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.