Please login to your online account to display your discounted pricing

3-Bromothiophenol, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Bromothiophenol
CAS 6320-01-0
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C6H5BrS
Linear Formula BrC6H4SH
Formula Weight 189.07
MDL Number MFCD00009603
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC303710050

  SDS
thermo fisher scientific
AC303710050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC303710010

  SDS
thermo fisher scientific
303710010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (GC)
Formula Weight 189.07
Physical Form Crystalline Powder
Quantity 5g
Color White to light beige
Density 1.632g/mL
CAS Min % 94.0
CAS Max % 100.0
Linear Formula BrC6H4SH
Synonym 3-Bromobenzenethiol
Infrared Spectrum Authentic
Chemical Name or Material 3-Bromothiophenol
CAS 6320-01-0
Assay Percent Range 95%
Molecular Formula C6H5BrS
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00009603
Packaging Glass bottle
Refractive Index 1.6320 to 1.6350 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic in contact with skin.
Causes skin irritation.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.