Please login to your online account to display your discounted pricing

3-Bromothiophene-2-carboxylic acid, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Bromothiophene-2-carboxylic acid
CAS 7311-64-0
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C5H3BrO2S
Formula Weight 207.05
MDL Number MFCD00052291
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW01838DA

  SDS
fisher scientific company
SEW01838DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.05
Quantity 1g
Assay 95%
CAS Min % 95.0
Packaging Amber Glass Bottle
Chemical Name or Material 3-Bromothiophene-2-carboxylic acid
Molecular Formula C5H3BrO2S
MDL Number MFCD00052291
CAS 7311-64-0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.