Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromothiophene-2-carboxaldehyde diethyl acetal, 97%

$94.47 - $323.96

Chemical Identifiers

Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde diethyl acetal
CAS 34042-95-0
Molecular Formula C9H13BrO2S
Formula Weight 265.17
MDL Number MFCD03426030
Beilstein 1284657
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1939703

  SDS
alfa aesar™
L1939703
1g Each for $94.47

AAL1939706

  SDS
alfa aesar™
L1939706
5g Each for $323.96
Description & Specifications

Specifications

Formula Weight 265.17
Molecular Formula C9H13BrO2S
Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde diethyl acetal
Beilstein 1284657
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Boiling Point 70°C (0.9mmHg)
Quantity 1g
MDL Number MFCD03426030
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Assay 97%
CAS 34042-95-0
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.