Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromothiophene-2-carboxaldehyde, 94%

$88.15 - $313.41

Chemical Identifiers

Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde
CAS 930-96-1
Molecular Formula C5H3BrOS
Formula Weight 191.05
MDL Number MFCD00126680
Beilstein 109757
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1938603

  SDS
alfa aesar™
L1938603
1g Each for $88.15

AAL1938606

  SDS
alfa aesar™
L1938606
5g Each for $313.41
Description & Specifications

Specifications

Formula Weight 191.05
Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde
Assay 94%
CAS 930-96-1
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Molecular Formula C5H3BrOS
MDL Number MFCD00126680
Beilstein 109757
Quantity 1g
Health Hazard 1 Warning
Boiling Point 110° to 113°C (10mmHg)

Applications
3-Bromothiophene-2-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.