Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Bromothiophene-2-carbonitrile, 97%

Chemical Identifiers

Chemical Name or Material 3-Bromothiophene-2-carbonitrile
CAS 18791-98-5
Molecular Formula C5H2BrNS
Formula Weight 188.05
MDL Number MFCD00068173
UN Number UN3439
Beilstein 1363930
Synonym 3-Bromo-2-cyanothiophene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0932803

  SDS
alfa aesar™
L0932803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL09328MD

  SDS
alfa aesar™
L09328MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.05
Chemical Name or Material 3-Bromothiophene-2-carbonitrile
Assay 97%
Molecular Formula C5H2BrNS
MDL Number MFCD00068173
UN Number UN3439
Synonym 3-Bromo-2-cyanothiophene
Health Hazard 2 GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
CAS 18791-98-5
Melting Point 46° to 51°C
Beilstein 1363930
Quantity 1g
Health Hazard 1 Danger
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H311-H302-H332-H315-H319
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.