missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromostyrene, 97%, stabilized, ACROS Organics™

Chemical Identifiers

CAS 2039-86-3
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H7Br
Linear Formula HC=CHC6H4Br
Formula Weight 183.05
Chemical Name or Material 3-Bromostyrene, Stabilized
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC306480010 SDS Thermo Fisher Scientific
AC306480010
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

2039-86-3
100.0
C8H7Br
183.05
≥96.0%
96.0
97%
HC=CHC6H4Br
3-Bromostyrene, Stabilized

Specifications

Liquid
1g
100.0
Authentic
97%
0.05% 3, 5-di-tert.-butylcatechol
3-Bromostyrene, Stabilized
183.05
Clear colorless to yellow
96.0
C8H7Br
2039-86-3
HC=CHC6H4Br
≥96.0%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.