missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Bromopyruvic acid hydrate, 98%, ACROS Organics™

$301.50 - $301.50

Chemical Identifiers

CAS 206860-50-6
Molecular Formula C3H3BrO3·xH2O
Molecular Weight (g/mol) 166.96
MDL Number MFCD00149107
InChI Key RQYDPLZNPJYABO-UHFFFAOYSA-N
Synonym 3-bromo-2-oxopropanoic acid hydrate, 3-bromo-2-oxopropanoic acid, hydrate, 3-bromo-2-oxopropanoic acid-water 1/1, 3-bromopyruvic acid hydrate, bromopyruvic acid hydrate, propanoic acid, 3-bromo-2-oxo-, monohydrate 9ci
PubChem CID 44118703
IUPAC Name 3-bromo-2-oxopropanoic acid;hydrate
SMILES C(C(=O)C(=O)O)Br.O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC325690250
SDS View Documents
Thermo Fisher Scientific
325690250
25g Glass bottle
Each for $301.50
 
Description

Description

Specifications

Chemical Identifiers

206860-50-6
166.96
RQYDPLZNPJYABO-UHFFFAOYSA-N
44118703
C(C(=O)C(=O)O)Br.O
C3H3BrO3·xH2O
MFCD00149107
3-bromo-2-oxopropanoic acid hydrate, 3-bromo-2-oxopropanoic acid, hydrate, 3-bromo-2-oxopropanoic acid-water 1/1, 3-bromopyruvic acid hydrate, bromopyruvic acid hydrate, propanoic acid, 3-bromo-2-oxo-, monohydrate 9ci
3-bromo-2-oxopropanoic acid;hydrate

Specifications

Authentic
Beige to Brown
25g
97.5
98%
MFCD00149107
Solubility in water: soluble
C(C(=O)C(=O)O)Br.O
166.96
166.96
98%
Glass bottle
79°C to 82°C
206860-50-6,1113-59-3
100.0
C3H3BrO3·xH2O
3-bromo-2-oxopropanoic acid hydrate, 3-bromo-2-oxopropanoic acid, hydrate, 3-bromo-2-oxopropanoic acid-water 1/1, 3-bromopyruvic acid hydrate, bromopyruvic acid hydrate, propanoic acid, 3-bromo-2-oxo-, monohydrate 9ci
RQYDPLZNPJYABO-UHFFFAOYSA-N
3-bromo-2-oxopropanoic acid;hydrate
44118703
Powder
3-Bromopyruvic acid hydrate, 98%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Danger

EINECSNumber : 214-206-5

RTECSNumber : UZ0840000

TSCA : TSCA

SDS